Assay Details
Target Gene Details
Entrez Gene ID: | 68401 |
Gene Name: | glucose 6 phosphatase, catalytic, 3 |
Gene Aliases: |
0710001K01Rik, AU019276, AU045429, AV128920, AW545836, UGRP |
Location: |
Chr.11:102189699-102194081 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G6pc3 | NM_175935.3 | 1 | 292 | NP_787949.2 |
| AK002966.1 | 2 | 156 | ||
| AK165395.1 | 1 | 146 | BAE38159.1 | |
| AY186239.1 | 1 | 138 | AAO39164.1 | |
| BC069959.1 | 1 | 292 | AAH69959.1 |
More Information
Set Membership: |
|

Genomic Map