Assay Details
Target Gene Details
Entrez Gene ID: | 22627 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon polypeptide |
Gene Aliases: |
AU019196 |
Location: |
Chr.11:75732887-75765841 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ywhae | NM_009536.4 | 6 | 1254 | NP_033562.3 |
| AK011612.1 | 3 | 623 | BAE43225.1 | |
| AK075995.1 | 6 | 1203 | BAC36106.1 | |
| AK135831.1 | 6 | 1182 | BAE22681.1 | |
| AK135867.1 | 6 | 1182 | BAE22705.1 | |
| AK169812.1 | 6 | 1209 | BAE41384.1 | |
| AK179881.1 | ||||
| AK180909.1 | ||||
| AK204067.1 | ||||
| AK204716.1 | ||||
| AK218111.1 | ||||
| AX775991.1 | 5 | 1022 | CAE11695.1 | |
| AX775993.1 | 4 | 815 | CAE11696.1 | |
| AY900661.1 | 1 | 140 | ||
| BC058686.1 | 6 | 1213 | AAH58686.1 | |
| D87663.1 | 6 | 1141 | BAA13424.1 | |
| Z19599.1 | 6 | 1196 | CAA79659.1 |
More Information
Set Membership: |
|

Genomic Map