Assay Details
Target Gene Details
Entrez Gene ID: | 108100 |
Gene Name: | brain-specific angiogenesis inhibitor 1-associated protein 2 |
Gene Aliases: |
IRSp53, R75030 |
Location: |
Chr.11:119942869-120006782 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baiap2 | NM_001037755.3 | 14 | 1921 | NP_001032844.2 |
| AK049469.1 | 14 | 1922 | BAC33764.1 | |
| AK170190.1 | 8 | 3463 | ||
| CO039918.1 | 1 | 163 |
More Information
Set Membership: |
|

Genomic Map