Assay Details
Target Gene Details
Entrez Gene ID: | 108100 |
Gene Name: | brain-specific angiogenesis inhibitor 1-associated protein 2 |
Gene Aliases: |
IRSp53, R75030 |
Location: |
Chr.11:119942869-120006782 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baiap2 | NM_001037754.3 | 14 | 2554 | NP_001032843.1 |
| NM_001037755.3 | NP_001032844.2 | |||
| NM_130862.4 | 14 | 2674 | NP_570932.2 | |
| AK049469.1 | BAC33764.1 | |||
| AK083201.1 | 1 | 769 | ||
| AK145924.1 | 14 | 2675 | BAE26755.1 | |
| AK158579.1 | 14 | 2696 | ||
| AK170190.1 | 7 | 2203 | ||
| AK186120.1 |
More Information
Set Membership: |
|

Genomic Map