Assay Details
Target Gene Details
Entrez Gene ID: | 68401 |
Gene Name: | glucose 6 phosphatase, catalytic, 3 |
Gene Aliases: |
0710001K01Rik, AU019276, AU045429, AV128920, AW545836, UGRP |
Location: |
Chr.11:102189699-102194081 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G6pc3 | NM_175935.3 | 6 | 1428 | NP_787949.2 |
| AK002966.1 | 7 | 1291 | ||
| AK165395.1 | 6 | 1282 | BAE38159.1 | |
| AK189516.1 | 1 | 68 | ||
| AY186239.1 | 6 | 1274 | AAO39164.1 | |
| BC069959.1 | 6 | 1428 | AAH69959.1 |
More Information
Set Membership: |
|

Genomic Map