Assay Details
Target Gene Details
Entrez Gene ID: | 14674 |
Gene Name: | guanine nucleotide binding protein, alpha 13 |
Gene Aliases: |
AU024132, AU043124, Galpha13 |
Location: |
Chr.11:109362783-109401369 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gna13 | NM_010303.3 | 4 | 3473 | NP_034433.3 |
| AK038290.1 | 1 | 558 | ||
| AK164985.1 | 1 | 651 |
More Information
Set Membership: |
|

Genomic Map