Assay Details
Target Gene Details
Entrez Gene ID: | 14534 |
Gene Name: | K(lysine) acetyltransferase 2A |
Gene Aliases: |
1110051E14Rik, AW212720, Gcn5, Gcn5l2, mmGCN5 |
Location: |
Chr.11:100704746-100712467 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat2a | NM_001038010.2 | 18 | 2929 | NP_001033099.1 |
| NM_020004.5 | 18 | 2932 | NP_064388.2 | |
| AK004229.1 | 1 | 360 | ||
| AK145205.1 | 16 | 2929 | BAE26296.1 | |
| AK147042.1 | 18 | 2877 | ||
| AK150225.1 | 18 | 2910 | BAE29392.1 | |
| AK154866.1 | 17 | 3007 | ||
| AK155791.1 | 18 | 2931 | ||
| AK155842.1 | 18 | 2930 | BAE33458.1 | |
| AK192506.1 | 1 | 183 | ||
| AK197611.1 | 1 | 183 | ||
| AK199655.1 | 1 | 183 | ||
| AV014576.1 | 1 | 152 | ||
| BC003983.1 | 3 | 727 | AAH03983.1 | |
| BC063752.1 | 18 | 2914 | AAH63752.1 |

Genomic Map