Assay Details
Target Gene Details
Entrez Gene ID: | 23989 |
Gene Name: | mediator complex subunit 24 |
Gene Aliases: |
911GSE, AU040102, AW547152, D11Ertd307e, DRIP100, Gse2, Pparb2, Pparbp2, R75526, Thrap4, Trap100 |
Location: |
Chr.11:98704591-98729475 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med24 | NM_011869.2 | 26 | 3053 | NP_035999.2 |
| AF126543.1 | 27 | 3131 | AAD42776.1 | |
| AF483498.1 | 25 | 2823 | AAL90772.1 | |
| AF483499.1 | 25 | 2823 | AAL90773.1 | |
| AK154618.1 | 26 | 3038 | BAE32717.1 | |
| AK191628.1 | ||||
| AK192379.1 | ||||
| AK209354.1 | ||||
| AK214599.1 | ||||
| AW541655.2 | 1 | 354 | ||
| BC005409.1 | 26 | 3075 | AAH05409.1 |
More Information
Set Membership: |
|

Genomic Map