Assay Details
Target Gene Details
Entrez Gene ID: | 192191 |
Gene Name: | mediator complex subunit 9 |
Gene Aliases: |
BC019367, Med25 |
Location: |
Chr.11:59948214-59963306 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med9 | NM_138675.3 | 2 | 1638 | NP_619616.2 |
| AK077295.1 | 2 | 1398 | BAC36735.2 | |
| AK147123.1 | 2 | 1630 | BAE27694.1 |
More Information
Set Membership: |
|

Genomic Map