Assay Details
Target Gene Details
Entrez Gene ID: | 338367 |
Gene Name: | myosin ID |
Gene Aliases: |
9930104H07Rik, AW544947, D11Ertd9e, myosin-1d |
Location: |
Chr.11:80482126-80780055 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myo1d | NM_177390.3 | 22 | 4974 | NP_796364.2 |
| AK148112.1 | 9 | 3093 | BAE28351.1 | |
| BC034071.1 | 8 | 2954 | AAH34071.1 |
More Information
Set Membership: |
|

Genomic Map