Assay Details
Target Gene Details
Entrez Gene ID: | 70802 |
Gene Name: | PWWP domain containing 2A |
Gene Aliases: |
4631424J17Rik, AI891583, AU024105, D930040F23Rik |
Location: |
Chr.11:43681082-43722633 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pwwp2a |

Genomic Map