Assay Details
Target Gene Details
Entrez Gene ID: | 70802 |
Gene Name: | PWWP domain containing 2A |
Gene Aliases: |
4631424J17Rik, AI891583, AU024105, D930040F23Rik |
Location: |
Chr.11:43681082-43722633 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pwwp2a | NM_001164231.1 | 2 | 1507 | NP_001157703.1 |
| NM_027557.1 | 2 | 1507 | NP_081833.1 | |
| AK014534.1 | 1 | 467 | BAB29417.1 | |
| AK171257.1 | 3 | 1700 | BAE42348.1 | |
| AK173305.1 | 2 | 1499 | BAD32583.1 |
More Information
Set Membership: |
|

Genomic Map