Assay Details
Target Gene Details
Entrez Gene ID: | 104910 |
Gene Name: | solute carrier family 25, member 47 |
Gene Aliases: |
AI132487, AI876593, HDMCP |
Location: |
Chr.12:108849823-108856815 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a47 | NM_001012310.2 | 5 | 482 | NP_001012310.2 |
| AK149369.1 | 5 | 488 | BAE28839.1 | |
| BC055027.1 | 5 | 475 | AAH55027.1 | |
| BC069939.1 | 5 | 471 | ||
| BC089616.1 | 5 | 482 | AAH89616.1 |
More Information
Set Membership: |
|

Genomic Map