Assay Details
Target Gene Details
Entrez Gene ID: | 69909 |
Gene Name: | RIKEN cDNA 2610027K06 gene |
Gene Aliases: |
- |
Location: |
Chr.11:85791660-85832388 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610027K06Rik | NR_077059.1 | |||
| AI425723.1 | ||||
| AK011568.1 | ||||
| AK085522.1 |
Target Gene Details
Entrez Gene ID: | 192197 |
Gene Name: | breast carcinoma amplified sequence 3 |
Gene Aliases: |
1500019F07Rik, 2610028P08Rik, AU021018, BC028339, K20D4, rudhira |
Location: |
Chr.11:85353116-85826058 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcas3 | NM_001166642.1 | 25 | 3457 | NP_001160114.1 |
| NM_001166643.1 | 24 | 3371 | NP_001160115.1 | |
| NM_138681.4 | 26 | 3502 | NP_619622.3 | |
| AK011603.1 | 5 | 1102 | ||
| AK032423.1 | 26 | 3453 | BAC27862.1 | |
| AK157222.1 | 25 | 3407 |
More Information
Set Membership: |
|

Genomic Map