Assay Details
Target Gene Details
Entrez Gene ID: | 14778 |
Gene Name: | glutathione peroxidase 3 |
Gene Aliases: |
AA960521, EGPx, GPx, GSHPx-3, GSHPx-P |
Location: |
Chr.11:54902854-54910382 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpx3 | NM_008161.3 | 5 | 1364 | NP_032187.2 |
| AA062309.1 | 1 | 182 | ||
| AI194448.1 | 1 | 135 | ||
| AK002219.1 | 5 | 1293 | BAC55243.1 | |
| AK002262.1 | 5 | 1293 | ||
| AK004942.1 | 5 | 1293 | BAC55250.1 | |
| AK146760.1 | 5 | 1293 | BAE27413.1 | |
| BC003339.1 | 5 | 1276 | AAH03339.1 | |
| BC037027.1 | 5 | 1311 | AAH37027.1 | |
| BC049235.1 | 5 | 1280 | AAH49235.1 | |
| BC061950.1 | 5 | 1268 | AAH61950.1 | |
| U13705.1 | 5 | 1268 | AAA62283.2 |
More Information
Set Membership: |
|

Genomic Map