Assay Details
Target Gene Details
Entrez Gene ID: | 71520 |
Gene Name: | GRB2-related adaptor protein |
Gene Aliases: |
8430435N19Rik, BB233594 |
Location: |
Chr.11:61653321-61672777 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grap | NM_027817.3 | 5 | 861 | NP_082093.1 |
| AK018457.1 | 5 | 862 | BAB31222.1 | |
| AK153888.1 | 5 | 918 | BAE32235.1 | |
| BC120674.1 | 5 | 856 | ||
| BC120676.1 | 5 | 856 |

Genomic Map