Assay Details
Target Gene Details
Entrez Gene ID: | 217127 |
Gene Name: | K(lysine) acetyltransferase 7 |
Gene Aliases: |
Hbo1, Hboa, Myst2 |
Location: |
Chr.11:95271853-95310246 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat7 | NM_001195003.1 | 15 | 2477 | NP_001181932.1 |
| NM_001195004.1 | 14 | 2387 | NP_001181933.1 | |
| NM_177619.3 | 13 | 2210 | NP_808287.1 | |
| AK147968.1 | 14 | 2204 | BAE28257.1 | |
| AK169458.1 | 11 | 2393 | ||
| AK170389.1 | 14 | 2334 | BAE41763.1 | |
| AK178421.1 | ||||
| AK179227.1 | ||||
| AK179739.1 | ||||
| BC048904.1 | 13 | 2165 | AAH48904.1 | |
| BC057102.1 | 14 | 2315 | AAH57102.1 |
More Information
Set Membership: |
|

Genomic Map