Assay Details
Target Gene Details
Entrez Gene ID: | 69900 |
Gene Name: | major facilitator superfamily domain containing 11 |
Gene Aliases: |
2600014M03Rik |
Location: |
Chr.11:116852283-116875660 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd11 |
Target Gene Details
Entrez Gene ID: | 20382 |
Gene Name: | serine/arginine-rich splicing factor 2 |
Gene Aliases: |
D11Wsu175e, MRF-1, Pr264, SC35, Sfrs10, Sfrs2 |
Location: |
Chr.11:116849897-116853104 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf2 | NM_011358.2 | 1 | 352 | NP_035488.1 |
| AF077858.1 | 1 | 340 | AAC71000.1 | |
| AK011528.1 | 1 | 349 | ||
| AK076448.1 | 1 | 350 | BAC36346.1 | |
| AK086103.1 | 1 | 350 | BAC39610.1 | |
| AK088042.1 | 1 | 350 | BAC40111.1 | |
| AK161873.1 | 1 | 348 | ||
| AK165670.1 | 1 | 354 | BAE38330.1 | |
| BC005493.1 | 1 | 330 | AAH05493.1 | |
| CJ043688.1 | 1 | 353 | ||
| L13610.1 | 1 | 325 | AAA39332.1 | |
| U14648.1 | 1 | 349 | AAA64595.1 |
More Information
Set Membership: |
|

Genomic Map