Assay Details
Target Gene Details
Entrez Gene ID: | 72258 |
Gene Name: | potassium channel, subfamily K, member 10 |
Gene Aliases: |
1700024D23Rik, 3010005K24Rik, Trek2 |
Location: |
Chr.12:98429432-98577940 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk10 | NM_029911.4 | 7 | 1347 | NP_084187.2 |
| AK031904.1 | 7 | 1278 | ||
| AK036066.1 | 7 | 1621 | BAC29295.1 | |
| AK052327.1 | 1 | 210 | BAC34939.1 | |
| AK082153.1 | 7 | 1348 | BAC38424.1 | |
| BC132487.1 | 7 | 1289 | ||
| BC137869.1 | 7 | 1289 | ||
| DQ185134.1 | 7 | 1244 | ABA28315.1 |
More Information
Set Membership: |
|

Genomic Map