Assay Details
Target Gene Details
Entrez Gene ID: | 15357 |
Gene Name: | 3-hydroxy-3-methylglutaryl-Coenzyme A reductase |
Gene Aliases: |
HMG-CoAR, Red |
Location: |
Chr.13:96648962-96670936 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgcr | NM_008255.2 | 20 | 3882 | NP_032281.2 |
| AK079302.1 | 8 | 2081 | BAC37602.1 | |
| AK159899.1 | 19 | 3770 | ||
| BC059873.1 | 11 | 2563 | AAH59873.1 | |
| BC085083.1 | 20 | 3794 | AAH85083.1 |
More Information
Set Membership: |
|

Genomic Map