Assay Details
Target Gene Details
Entrez Gene ID: | 320174 |
Gene Name: | RIKEN cDNA A830082K12 gene |
Gene Aliases: |
- |
Location: |
Chr.13:78198017-78236564 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A830082K12Rik | NR_045195.1 | |||
| AK082108.1 |
Target Gene Details
Entrez Gene ID: | 13865 |
Gene Name: | nuclear receptor subfamily 2, group F, member 1 |
Gene Aliases: |
COUP-TF1, COUP-TFI, COUPTFA, EAR3, Erbal3, SVP44, Tcfcoup1 |
Location: |
Chr.13:78188971-78199172 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr2f1 | NM_010151.2 | 1 | 435 | NP_034281.2 |
| BC108408.1 | 1 | 324 | AAI08409.1 | |
| U07625.1 | AAA19853.1 | |||
| X74134.1 | 1 | 326 | CAA52231.1 |
More Information
Set Membership: |
|

Genomic Map