Assay Details
Target Gene Details
Entrez Gene ID: | 192119 |
Gene Name: | dicer 1, ribonuclease type III |
Gene Aliases: |
1110006F08Rik, D12Ertd7e, mKIAA0928 |
Location: |
Chr.12:104687740-104751952 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dicer1 | NM_148948.2 | 27 | 9182 | NP_683750.2 |
| AK003494.1 |

Genomic Map