Assay Details
Target Gene Details
Entrez Gene ID: | 13865 |
Gene Name: | nuclear receptor subfamily 2, group F, member 1 |
Gene Aliases: |
COUP-TF1, COUP-TFI, COUPTFA, EAR3, Erbal3, SVP44, Tcfcoup1 |
Location: |
Chr.13:78188971-78199172 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr2f1 | NM_010151.2 | 3 | 1740 | NP_034281.2 |
| AK148880.1 | 3 | 2077 | BAE28687.1 | |
| BC108408.1 | 3 | 1629 | AAI08409.1 | |
| U07625.1 | 3 | 1342 | AAA19853.1 | |
| X74134.1 | 3 | 1640 | CAA52231.1 |
More Information
Set Membership: |
|

Genomic Map