Assay Details
Target Gene Details
Entrez Gene ID: | 69480 |
Gene Name: | tetratricopeptide repeat domain 9 |
Gene Aliases: |
1700029M07Rik, AI429215, AI853559 |
Location: |
Chr.12:81629771-81664941 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc9 | NM_001033149.2 | 3 | 1327 | NP_001028321.1 |
| AK006508.1 | 1 | 439 | ||
| AK133456.1 | 3 | 1328 | BAE21667.1 | |
| AK161499.1 | 3 | 1127 | BAE36425.1 | |
| BC028891.1 | 3 | 1411 | AAH28891.1 |
More Information
Set Membership: |
|

Genomic Map