Assay Details
Target Gene Details
Entrez Gene ID: | 380840 |
Gene Name: | LYR motif containing 4 |
Gene Aliases: |
BC034664, Gm903 |
Location: |
Chr.13:35978797-36117357 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lyrm4 | NM_201358.2 | NP_958746.1 | ||
| AK050553.1 | BAC34321.1 | |||
| BC034664.1 | AAH34664.1 | |||
| BC080772.1 | AAH80772.1 |
More Information
Set Membership: |
|

Genomic Map