Assay Details
Target Gene Details
Entrez Gene ID: | 66222 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 1a |
Gene Aliases: |
1190005M04Rik, AI325983, EI, EIA, ELANH2, LEI, M/NEI, MNEI, PI2 |
Location: |
Chr.13:32842092-32851185 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb1a | NM_025429.2 | 7 | 1358 | NP_079705.2 |
| AK004500.1 | 7 | 1359 | BAB23335.1 | |
| AK160167.1 | 7 | 1354 | BAE35668.1 | |
| AK160456.1 | 1 | 326 | BAE35797.1 | |
| BC104333.2 | AAI04334.1 |
More Information
Set Membership: |
|

Genomic Map