Assay Details
Target Gene Details
Entrez Gene ID: | 26932 |
Gene Name: | protein phosphatase 2, regulatory subunit B', epsilon |
Gene Aliases: |
4633401M22Rik, AI449017, B56beta |
Location: |
Chr.12:75449463-75596250 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r5e | NM_012024.2 | 1 | 382 | NP_036154.1 |
| AK053228.1 | 1 | 428 | ||
| AK078894.1 | 1 | 412 | ||
| AK088366.1 | 1 | 352 | BAC40306.1 | |
| AK132432.1 | 1 | 415 | BAE21166.1 | |
| AK133824.1 | 1 | 427 | BAE21863.1 | |
| AK220163.1 | 1 | 375 | BAD90335.1 | |
| BC085149.1 | 1 | 382 | AAH85149.1 |
More Information
Set Membership: |
|

Genomic Map