Assay Details
Target Gene Details
Entrez Gene ID: | 214663 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, palmitoylcarnitine transporter), member 29 |
Gene Aliases: |
C030003J19Rik, CACL, mCACL |
Location: |
Chr.12:108825878-108835876 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a29 | NM_181328.3 | 4 | 1153 | NP_851845.1 |
| AK047628.1 | 4 | 1154 | BAC33105.1 | |
| AK087057.1 | 4 | 1140 | ||
| AK148350.1 | 4 | 1131 | ||
| AK160906.1 | 4 | 1102 | ||
| AK177416.1 | 1 | 281 | ||
| BC006711.1 | 4 | 1026 | AAH06711.1 |
More Information
Set Membership: |
|

Genomic Map