Assay Details
Target Gene Details
Entrez Gene ID: | 26950 |
Gene Name: | visinin-like 1 |
Gene Aliases: |
VILIP, Vnsl1 |
Location: |
Chr.12:11325245-11436649 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vsnl1 | NM_012038.4 | 1 | 256 | NP_036168.1 |
| AJ489331.1 | 1 | 220 | CAD33793.1 | |
| AK144454.1 | 1 | 186 | BAE25898.1 | |
| AK160224.1 | 1 | 188 | BAE35699.1 | |
| BC046226.1 | 1 | 161 | AAH46226.1 | |
| BY264558.1 | 1 | 262 |

Genomic Map