Assay Details
Target Gene Details
Entrez Gene ID: | 26950 |
Gene Name: | visinin-like 1 |
Gene Aliases: |
VILIP, Vnsl1 |
Location: |
Chr.12:11325245-11436649 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vsnl1 | NM_012038.4 | 4 | 1306 | NP_036168.1 |
| AJ489331.1 | 4 | 1270 | CAD33793.1 | |
| AK005091.2 | 3 | 868 | ||
| AK144454.1 | 4 | 1236 | BAE25898.1 | |
| AY101375.1 | 3 | 1008 | AAM48292.1 |

Genomic Map