Assay Details
Target Gene Details
Entrez Gene ID: | 218544 |
Gene Name: | small glutamine-rich tetratricopeptide repeat (TPR)-containing, beta |
Gene Aliases: |
C630001O05Rik |
Location: |
Chr.13:104109724-104142492 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgtb | NM_144838.1 | 11 | 1569 | NP_659087.1 |
| AK049820.1 | 11 | 1607 | BAC33934.1 | |
| AK082080.1 | 11 | 1459 | BAC38406.1 | |
| AK192921.1 | ||||
| AK203597.1 | ||||
| AK209162.1 | ||||
| BC017611.1 | 11 | 1569 | AAH17611.1 |

Genomic Map