Assay Details
Target Gene Details
Entrez Gene ID: | 211948 |
Gene Name: | phosphodiesterase 12 |
Gene Aliases: |
2'-PDE, E430028B21Rik |
Location: |
Chr.14:26664117-26669846 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde12 | NM_178668.3 | 3 | 3231 | NP_848783.3 |
| AK143567.1 | 3 | 3058 | ||
| AK161083.1 | 3 | 3271 | BAE36189.1 | |
| BC064450.1 | 3 | 3231 | AAH64450.1 |
More Information
Set Membership: |
|

Genomic Map