Assay Details
Target Gene Details
Entrez Gene ID: | 320808 |
Gene Name: | DDB1 and CUL4 associated factor 5 |
Gene Aliases: |
9430020B07Rik, AI325023, AI430035, AI848110, AW124614, BCRG2, BCRP2, Wdr22 |
Location: |
Chr.12:80335847-80436601 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf5 | NM_177267.4 | 9 | 3070 | NP_796241.3 |
| AK034653.1 | 1 | 1652 | BAC28784.1 | |
| AK077889.1 | 1 | 569 | BAC37050.1 | |
| AK122561.1 | 9 | 2907 | BAC65843.1 | |
| AK137579.1 | 9 | 3070 | BAE23416.1 | |
| AK165096.1 | 9 | 3054 | ||
| BC030857.1 | 1 | 1283 | AAH30857.1 | |
| BC042567.1 | 9 | 2744 | AAH42567.1 | |
| BC046813.1 | 9 | 3008 | AAH46813.2 |
More Information
Set Membership: |
|

Genomic Map