Assay Details
Target Gene Details
Entrez Gene ID: | 56410 |
Gene Name: | cerebellin 3 precursor protein |
Gene Aliases: |
- |
Location: |
Chr.14:55878920-55884256 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cbln3 | NM_019820.3 | 3 | 1179 | NP_062794.1 |
| AF218379.1 | 4 | 1035 | AAF32314.1 | |
| AK081085.1 | 3 | 1155 | BAC38129.1 | |
| BC065108.1 | 3 | 1179 | AAH65108.1 |

Genomic Map