Assay Details
Target Gene Details
Entrez Gene ID: | 100303738 |
Gene Name: | RIKEN cDNA D030025P21 gene |
Gene Aliases: |
- |
Location: |
Chr.12:84875802-84879755 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D030025P21Rik | NR_028577.1 | 1 | 324 | |
| AK144083.1 | 1 | 325 |
Target Gene Details
Entrez Gene ID: | 16997 |
Gene Name: | latent transforming growth factor beta binding protein 2 |
Gene Aliases: |
AW208642 |
Location: |
Chr.12:84783212-84877266 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltbp2 | NM_013589.3 | 1 | 371 | NP_038617.3 |
| AF004874.1 | 1 | 408 | AAB61611.1 | |
| AK052980.1 | 1 | 368 | BAC35229.1 | |
| BB842382.1 | ||||
| BC119785.1 | 1 | 355 |
More Information
Set Membership: |
|

Genomic Map