Assay Details
Target Gene Details
Entrez Gene ID: | 16997 |
Gene Name: | latent transforming growth factor beta binding protein 2 |
Gene Aliases: |
AW208642 |
Location: |
Chr.12:84783212-84877266 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltbp2 | NM_013589.3 | 33 | 5085 | NP_038617.3 |
| AF004874.1 | 34 | 5264 | AAB61611.1 | |
| BC119785.1 | 33 | 5069 |
More Information
Set Membership: |
|

Genomic Map