Assay Details
Target Gene Details
Entrez Gene ID: | 24000 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 21 |
Gene Aliases: |
PTPD1, PTPRL10 |
Location: |
Chr.12:98676741-98737618 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn21 | NM_001146199.1 | 19 | 4926 | NP_001139671.1 |
| NM_011877.2 | 19 | 5061 | NP_036007.2 | |
| D37801.1 | 19 | 5010 | BAA07053.1 | |
| D83072.1 | 6 | 2322 | BAA19740.1 |
More Information
Set Membership: |
|

Genomic Map