Assay Details
Target Gene Details
Entrez Gene ID: | 58248 |
Gene Name: | RIKEN cDNA 1700123O20 gene |
Gene Aliases: |
MNCb-2990 |
Location: |
Chr.14:54686171-54690742 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700123O20Rik | NM_021437.2 | 1 | 787 | NP_067412.1 |
| AB041657.1 | 1 | 742 | BAA95104.1 | |
| AK007261.1 | 1 | 188 | BAB24921.1 | |
| AK030845.1 | 1 | 743 | ||
| AK167989.1 | BAE39980.1 | |||
| AK170890.1 | 1 | 121 | BAE42097.1 |
Target Gene Details
Entrez Gene ID: | 56215 |
Gene Name: | apoptotic chromatin condensation inducer 1 |
Gene Aliases: |
2610036I19Rik, 2610510L13Rik, Acinus, Acn, C79325, acinusL, acinusS, mKIAA0670 |
Location: |
Chr.14:54642161-54686931 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acin1 | NM_001085473.2 | NP_001078942.2 | ||
| NM_023190.3 | NP_075679.2 | |||
| AF168782.1 | AAF89661.1 | |||
| CF165831.1 |

Genomic Map