Assay Details
Target Gene Details
Entrez Gene ID: | 66975 |
Gene Name: | trafficking protein particle complex 13 |
Gene Aliases: |
2410002O22Rik, 2610524F24Rik, AI451892, AU017229 |
Location: |
Chr.13:104142153-104178703 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc13 | NM_001093759.1 | 1 | 234 | NP_001087228.1 |
| NM_001093760.1 | 1 | 234 | NP_001087229.1 | |
| NM_025879.2 | 1 | 234 | NP_080155.3 | |
| NR_003546.1 | 1 | 234 | ||
| AK010346.1 | 1 | 215 | BAB26869.2 | |
| AK012141.1 | 1 | 209 | BAB28057.1 | |
| AK013956.1 | 1 | 192 | BAB29083.2 | |
| AK084373.1 | 1 | 184 | BAC39168.1 | |
| AK149832.1 | 1 | 237 | BAE29111.1 | |
| AK163342.1 | 1 | 222 | BAE37309.1 | |
| AK167749.1 | 1 | 194 | BAE39785.1 | |
| BC021756.1 | 1 | 175 |

Genomic Map