Assay Details
Target Gene Details
Entrez Gene ID: | 268697 |
Gene Name: | cyclin B1 |
Gene Aliases: |
Ccnb1-rs1, Ccnb1-rs13, CycB1, Cycb-4, Cycb-5, Cycb1-rs1 |
Location: |
Chr.13:100778732-100786543 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnb1 | NM_172301.3 | 9 | 2148 | NP_758505.2 |
| AK088816.1 | 2 | 2311 | ||
| AK136259.1 | 9 | 2162 | BAE22901.1 | |
| AK143597.1 | 9 | 2104 | BAE25456.1 | |
| AK166686.1 | 9 | 2162 | BAE38945.1 | |
| AK167950.1 | 9 | 2161 | ||
| AK168502.1 | 9 | 2162 | BAE40387.1 | |
| AK168993.1 | 9 | 2159 | BAE40792.1 | |
| BC011478.1 | 9 | 2148 | AAH11478.1 | |
| BC080202.1 | 8 | 1935 | AAH80202.1 | |
| S43105.1 | 9 | 2186 | AAB22970.1 | |
| X64713.1 | 9 | 2139 | CAA45968.1 |
More Information
Set Membership: |
|

Genomic Map