Assay Details
Target Gene Details
Entrez Gene ID: | 268564 |
Gene Name: | zinc finger and BTB domain containing 1 |
Gene Aliases: |
9430077A12, AI256369, Bsg2, C430003J21Rik |
Location: |
Chr.12:76370192-76396955 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb1 | NM_178744.2 | 2 | 1545 | NP_848859.1 |
| AK029762.1 | 2 | 1293 | BAC26603.1 | |
| AK035036.1 | 2 | 1625 | BAC28921.1 | |
| AK049397.1 | 2 | 1545 | BAC33733.1 | |
| AK131985.1 | 2 | 1228 | BAE20921.1 | |
| AY611626.1 | 2 | 1545 | AAU14845.1 | |
| BC012239.1 | 2 | 1209 | AAH12239.1 |
More Information
Set Membership: |
|

Genomic Map