Assay Details
Target Gene Details
Entrez Gene ID: | 67842 |
Gene Name: | NOP9 nucleolar protein |
Gene Aliases: |
2610027L16Rik |
Location: |
Chr.14:55745693-55755634 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nop9 | NM_026403.3 | 2 | 483 | NP_080679.3 |
| AK011570.1 | 2 | 473 | BAB27705.1 | |
| AK015585.1 | 2 | 233 | BAB29900.1 | |
| AK032873.1 | 2 | 467 | BAC28064.1 | |
| AK037831.1 | 2 | 471 | BAC29882.1 | |
| AK050912.1 | 2 | 473 | BAC34455.1 | |
| AK083861.1 | 2 | 352 | BAC39042.1 | |
| AK139689.1 | 2 | 433 | ||
| AK146020.1 | 2 | 438 | BAE26838.1 | |
| AK146687.1 | 2 | 454 | ||
| AK152732.1 | 2 | 457 | BAE31453.1 | |
| AK154387.1 | 2 | 475 | BAE32553.1 | |
| AK159798.1 | 2 | 457 | BAE35379.1 | |
| AK185037.1 | ||||
| AK219900.1 | ||||
| BC018523.1 | 2 | 463 | AAH18523.1 | |
| CJ098414.1 |
More Information
Set Membership: |
|

Genomic Map