Assay Details
Target Gene Details
Entrez Gene ID: | 18191 |
Gene Name: | neurexin III |
Gene Aliases: |
- |
Location: |
Chr.12:88722865-90334935 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrxn3 | NM_001198587.3 | NP_001185516.2 | ||
| AK078216.1 | ||||
| AK082055.1 | ||||
| AK129205.1 | BAC98015.2 | |||
| BC060719.1 | AAH60719.1 |
More Information
Set Membership: |
|

Genomic Map