Assay Details
Target Gene Details
Entrez Gene ID: | 20773 |
Gene Name: | serine palmitoyltransferase, long chain base subunit 2 |
Gene Aliases: |
AI173915, LCB2, mKIAA0526 |
Location: |
Chr.12:87307889-87388230 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptlc2 | NM_011479.3 | 12 | 2687 | NP_035609.1 |
| AK042149.1 | 2 | 1201 | BAC31185.1 | |
| AK044917.1 | 12 | 2688 | BAC32140.1 | |
| AK080181.1 | 6 | 2885 | BAC37842.1 |
More Information
Set Membership: |
|

Genomic Map