Assay Details
Target Gene Details
Entrez Gene ID: | 18708 |
Gene Name: | phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha) |
Gene Aliases: |
PI3K, p50alpha, p55alpha, p85alpha |
Location: |
Chr.13:101680761-101768217 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3r1 | NM_001024955.2 | 10 | 4394 | NP_001020126.1 |
| NM_001077495.2 | 16 | 5483 | NP_001070963.1 | |
| AK049798.1 | 1 | 1329 | BAC33923.1 | |
| AK138868.1 | 1 | 1303 | BAE23803.1 |
More Information
Set Membership: |
|

Genomic Map