Assay Details
Target Gene Details
Entrez Gene ID: | 217721 |
Gene Name: | major facilitator superfamily domain containing 7C |
Gene Aliases: |
CCT, Flvcr2 |
Location: |
Chr.12:85746539-85813585 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd7c | NM_145447.2 | 10 | 2153 | NP_663422.1 |
| AK145696.1 | 10 | 2110 | BAE26594.1 | |
| AY260574.1 | 10 | 2153 | AAP86635.1 | |
| AY260575.1 | 10 | 2153 | AAP86636.1 | |
| AY260576.1 | 10 | 2153 | AAP86637.1 | |
| BC011209.1 | 10 | 2153 | AAH11209.1 |
More Information
Set Membership: |
|

Genomic Map