Assay Details
Target Gene Details
Entrez Gene ID: | 73274 |
Gene Name: | GC-rich promoter binding protein 1 |
Gene Aliases: |
1700034P14Rik, AU019836, D230035M11Rik, Gpbp, mGPBP |
Location: |
Chr.13:111425680-111490123 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpbp1 | NM_001122963.1 | 12 | 3401 | NP_001116435.1 |
| NM_028487.4 | 13 | 3461 | NP_082763.3 | |
| AY382529.1 | 13 | 3259 | AAQ88446.1 | |
| CF582635.1 | 1 | 194 |

Genomic Map