Assay Details
Target Gene Details
Entrez Gene ID: | 212937 |
Gene Name: | TRAF-interacting protein with forkhead-associated domain, family member B |
Gene Aliases: |
- |
Location: |
Chr.13:56173702-56179018 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tifab | NM_001168615.1 | 1 | 210 | NP_001162086.1 |
| NM_145976.4 | 1 | 210 | NP_666088.1 | |
| AK036643.1 | 1 | 127 | BAC29516.1 | |
| AK170349.1 | BAE41737.1 | |||
| BC031130.1 | AAH31130.1 | |||
| BY209244.1 | 1 | 210 |

Genomic Map