Assay Details
Target Gene Details
Entrez Gene ID: | 212937 |
Gene Name: | TRAF-interacting protein with forkhead-associated domain, family member B |
Gene Aliases: |
- |
Location: |
Chr.13:56173702-56179018 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tifab | NM_001168615.1 | 3 | 1055 | NP_001162086.1 |
| NM_145976.4 | 3 | 991 | NP_666088.1 | |
| AK036643.1 | 3 | 908 | BAC29516.1 | |
| AK153654.1 | 3 | 787 | BAE32135.1 | |
| AK163959.1 | ||||
| AK170349.1 | 3 | 836 | BAE41737.1 | |
| AK220206.1 | BAD90131.1 | |||
| BC005528.1 | 3 | 689 | ||
| BC027057.1 | 2 | 574 | AAH27057.1 | |
| BC031130.1 | 3 | 823 | AAH31130.1 |

Genomic Map