Assay Details
Target Gene Details
Entrez Gene ID: | 71037 |
Gene Name: | protease, serine 55 |
Gene Aliases: |
4933401F05Rik |
Location: |
Chr.14:64075435-64085433 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss55 | NM_001081063.1 | 5 | 964 | NP_001074532.1 |
| AK016601.1 | 4 | 990 | ||
| BC115562.2 | 5 | 965 |
More Information
Set Membership: |
|

Genomic Map